missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurantoin, MedChemExpress

Product Code. 16492009
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492009 1 g 1g
16490728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492009 Supplier Medchem Express Supplier No. HYA0090
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nitrofurantoin is a potent and orally active broad-spectrum beta-lactamase antimicrobial agent. Nitrofurantoin acts as an antibiotic and can be used for the study of urinary tract infections (UTIs), including cystitis and kidney infections.

Nitrofurantoin is a potent and orally active broad-spectrum beta-lactamase antimicrobial agent. Nitrofurantoin acts as an antibiotic and can be used for the study of urinary tract infections (UTIs), including cystitis and kidney infections.

Specifications

Chemical Name or Material Nitrofurantoin
Physical Form Solid
CAS 67-20-9
Color Light Yellow to Yellow
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C8H6N4O5
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (419.89 mM; Need ultrasonic)
SMILES O=C1NC(CN1/N=C/C2=CC=C([N+]([O-])=O)O2)=O
Molecular Weight (g/mol) 238.16
Formula Weight 238.16
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.