missing translation for 'onlineSavingsMsg'
Learn More

Nitrogen Check Standard: N @ 1 ppm in Isooctane, VHG™

Product Code. 17904176
Change view
Click to view available options
Quantity:
5 x 2 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904176 5 x 2 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17904176 Supplier VHG Labs Supplier No. ALKKTNISOV51SGL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Nitrogen Check Standard: N @ 1 ppm in Isooctane
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitrogen
CAS 7727-37-9
Analyte or Component Names Nitrogen
Shipping Condition Room Temperature
Molecular Formula N2
Concentration 1 ppm
Synonym Nitrogen, Diatomic nitrogen, Dinitrogen, Molecular nitrogen, Nitrogen (N2), Nitrogen gas, Nitrogen nutrition (plant), Nitrogen-14
InChI Formula InChI=1S/N2/c1-2
SMILES N#N
IUPAC Name molecular nitrogen
Solvent or Matrix Isooctane
Molecular Weight (g/mol) 28.01
Quantity 5 x 2 mL
Formula Weight 28.0061 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.