missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKG2D/CD314 Antibody (6E5A7), Alexa Fluor™ 594 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20156AF594
This item is not returnable.
View return policy
Description
NKG2D/CD314 Monoclonal antibody specifically detects NKG2D/CD314 in Human samples. It is validated for Flow Cytometry
Specifications
| NKG2D/CD314 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence | |
| The original antibody was generated by immunizing BALB/c mice with soluble NKG2-D fragments containing amino acids from 64-462 and 123-462. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Neuroscience, Signal Transduction, Stem Cells | |
| 22914 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Flow Cytometry | |
| 6E5A7 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction