missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKG2D/CD314 Antibody (6E5A7) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
244.00 € - 635.00 €
Specifications
| Antigen | NKG2D/CD314 |
|---|---|
| Clone | 6E5A7 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611734
|
Novus Biologicals
NBP3-20156-0.025mg |
0.025 mg |
244.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679803
|
Novus Biologicals
NBP3-20156-0.2mg |
0.2 mg |
635.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKG2D/CD314 Monoclonal antibody specifically detects NKG2D/CD314 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, NeutralizationSpecifications
| NKG2D/CD314 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence | |
| The original antibody was generated by immunizing BALB/c mice with soluble NKG2-D fragments containing amino acids from 64-462 and 123-462. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| 6E5A7 | |
| Western Blot, Flow Cytometry, Immunofluorescence, Neutralization | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology, Neuroscience, Signal Transduction, Stem Cells | |
| PBS | |
| 22914 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title