missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKR-P1 Antibody (3.2.3) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22437
This item is not returnable.
View return policy
Description
NKR-P1 Monoclonal specifically detects NKR-P1 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| NKR-P1 | |
| Monoclonal | |
| 0.51 mg/mL | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500, CyTOF-ready | |
| KLRB1 | |
| Rat 24.5 kD cell surface protein known as NKR-P1 | |
| 500 μg | |
| Primary | |
| This antibody reacts with Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| 3.2.3 | |
| Unconjugated | |
| killer cell lectin-like receptor subfamily B, member 1, Klrb1, Klrb1 killer cell lectin-like receptor subfamily B, member 1, Klrb1d, Klrb1f, Klrb1g, Klrb6, Nkr-p1e, Nkrp-1e, NKR-P1G | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 689817 | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction