missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NM23-H2/NME2 Mouse anti-Human, Clone: 2000, Novus Biologicals™
Mouse Monoclonal Antibody
294.00 € - 453.00 €
Specifications
| Antigen | NM23-H2/NME2 |
|---|---|
| Clone | 2000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18284133
|
Novus Biologicals
NBP2-60241-50UL |
50 μL |
294.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18238411
|
Novus Biologicals
NBP2-60241-100UL |
100 μL |
453.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NM23-H2/NME2 Monoclonal specifically detects NM23-H2/NME2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| NM23-H2/NME2 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 4831 | |
| Recombinant human NME2 (1-152aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 2000 | |
| Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in | |
| NME2 | |
| IgG1 κ | |
| Protein A purified |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title