missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR1 Antibody (S08-4B6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19718-100ul
This item is not returnable.
View return policy
Description
NMDAR1 Monoclonal antibody specifically detects NMDAR1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NMDAR1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S08-4B6 | |
| Unconjugated | |
| glutamate [NMDA] receptor subunit zeta-1, glutamate receptor, ionotropic, N-methyl D-aspartate 1, NMDA1, NMDAR1glutamate [NMDA] receptor subunit zeta 1, NMD-R1, N-methyl-D-aspartate receptor channel, subunit zeta-1, N-methyl-D-aspartate receptor subunit NR1, NR1 | |
| A synthetic peptide of human NMDAR1 (Uniprot # Q05586) | |
| 100 μg | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific | |
| 2902 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction