missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR2D Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94573-0.1ml
This item is not returnable.
View return policy
Description
NMDAR2D Polyclonal antibody specifically detects NMDAR2D in Human, Mouse samples. It is validated for Western Blot
Specifications
| NMDAR2D | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EB11estrogen receptor binding CpG island, glutamate receptor, ionotropic, N-methyl D-aspartate 2D, NMDA epsilon 4, NMDAR2DN-methyl-d-aspartate receptor subunit 2D, N-methyl D-aspartate receptor subtype 2D, NR2Dglutamate [NMDA] receptor subunit epsilon-4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 330-480 of human GRIN2D (NP_000827.2). GAQALLRDYGFLPELGHDCRAQNRTHRGESLHRYFMNITWDNRDYSFNEDGFLVNPSLVVISLTRDRTWEVVGSWEQQTLRLKYPLWSRYGRFLQPVDDTQHLTVATLEERPFVIVEPADPISGTCIRDSVPCRSQLNRTHSPPPDAPRPE | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 2906 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction