missing translation for 'onlineSavingsMsg'
Learn More

NME1 Monoclonal Antibody (OTI1A12), TrueMAB™, OriGene

Product Code. 17549582
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17549582 30 μL 30µL
17529582 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17549582 Supplier Origene Technologies Supplier No. TA801259S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NME1 Monoclonal antibody specifically detects NME1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen NME1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1A12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NME1
Gene Accession No. P15531
Gene Alias AL024257; AWD; expressed in non-metastatic cells 1; expressed in non-metastatic cells 1 protein; expressed in non-metastatic cells 1 protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 1, protein; expressed in non-metastatic cells 1, protein (NM23A) (nucleoside diphosphate kinase); GAAD; Granzyme A activated DNase (GAAD); Granzyme A-activated DNase; GZMA activated DNase; included; metastasis inhibition factor NM23; NB; NBR-A; NBR-B; NBS; NDK A; NDK A 1; NDK A 2; NDK NBR-A; NDK NBR-B; NDKA; NDP kinase A; NDP kinase A 1; NDP kinase A 2; NDP kinase beta; NDP kinase NBR-A; NDPKA; NDPK-A; NM23; NM23A; NM23-C1; NM23-H1; NM23H1B; nm23-M1; NME/NM23 nucleoside diphosphate kinase 1; Nme1; NME1-1; NME1-NME2 spliced read-through transcript; non-metastatic cells 1, protein (NM23A) expressed in; nucleoside diphosphate kinase A; nucleoside diphosphate kinase A 1; nucleoside diphosphate kinase A 2; nucleoside diphosphate kinase NBR-B; nucleoside-diphosphate kinase 1; nucleoside-diphosphate kinase NBR-A; nucleotide diphosphate kinase; nucloside diphosphate kinase; tumor metastatic process-associated protein
Host Species Mouse
Immunogen Full length human recombinant protein of human NME1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4830
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.