missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NNMT Mouse anti-Human, Mouse, Clone: OTI3D8, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502624
This item is not returnable.
View return policy
Description
N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver. This gene encodes the protein responsible for this enzymatic activity which uses S-adenosyl methionine as the methyl donor.Specifications
| NNMT | |
| Monoclonal | |
| Unconjugated | |
| NNMT | |
| nicotinamide N-methyltransferase | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 18113, 4837 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D8 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O55239, P40261 | |
| NNMT | |
| Full length human recombit protein of human NNMT produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction