missing translation for 'onlineSavingsMsg'
Learn More

NO3- f(N)-NITRATE solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
BIOPHARMA

Product Code. 12922165 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12922165 500 mL Bottle 500mL
12902165 100 mL Bottle 100mL
12912165 250 mL Bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12922165 Supplier Fisher Chemical Supplier No. A 031 500
  • ONLINE EXCLUSIVE PRICE
    126.00 €
    124.66 € / 500mL
    Save 1.34 €
    1% Off
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 14797-55-8
Molecular Formula NO3-
Molecular Weight (g/mol) 62.004
InChI Key NHNBFGGVMKEFGY-UHFFFAOYSA-N
PubChem CID 943
ChEBI CHEBI:17632
IUPAC Name nitrate
SMILES [N+](=O)([O-])[O-]

Specifications

Packaging Bottle
Physical Form Liquid
Concentration 1 g/L ±0.002
Type Single-component Standard Solution
Solvent or Matrix Water
Quantity 500 mL
Chemical Name or Material Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.