missing translation for 'onlineSavingsMsg'
Learn More

Nobiletin, MedChemExpress

Product Code. 16498878
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498878 10 mg 10mg
16469006 100 mg 100mg
16404777 50 mg 50mg
16401328 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498878 Supplier Medchem Express Supplier No. HYN0155
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nobiletin is a poly-methoxylated flavone from the citrus peel that improves memory loss. Nobiletin is a retinoid acid receptor-related orphan receptors (RORs) agonist. Nobiletin can reduce reactive oxygen species (ROS) levels in differentiated C2C12 myotubes and has anti-inflammation and anti-cancer properties, including anti-angiogenesis, anti-proliferation, anti-metastasis and induced apoptosis.

Nobiletin is a poly-methoxylated flavone from the citrus peel that improves memory loss. Nobiletin is a retinoid acid receptor-related orphan receptors (RORs) agonist. Nobiletin can reduce reactive oxygen species (ROS) levels in differentiated C2C12 myotubes and has anti-inflammation and anti-cancer properties, including anti-angiogenesis, anti-proliferation, anti-metastasis and induced apoptosis.

Specifications

Chemical Name or Material Nobiletin
CAS 478-01-3
Color White
Purity Grade Notes Research
Molecular Formula C21H22O8
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (124.26 mM; Need ultrasonic)
SMILES O=C1C=C(C2=CC=C(OC)C(OC)=C2)OC3=C(OC)C(OC)=C(OC)C(OC)=C13
Molecular Weight (g/mol) 402.39
Formula Weight 402.39
Percent Purity 98.19%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.