missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOC4L Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93101-0.02ml
This item is not returnable.
View return policy
Description
NOC4L Polyclonal antibody specifically detects NOC4L in Human samples. It is validated for Western Blot
Specifications
| NOC4L | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| MGC3162, NET49, NOC4, NOC4 protein homolog, NOC4-like protein, nucleolar complex associated 4 homolog (S. cerevisiae), nucleolar complex protein 4 homolog, Nucleolar complex-associated protein 4-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human NOC4L (NP_076983.1). MEREPGAAGVRRALGRRLEAVLASRSEANAVFDILAVLQSEDQEEIQEAVRTCSRLFGALLERGELFVGQLPSEEMVMTGSQGATRKYKVWMRHRYHSCCNRLGELLGHPSFQVKELALSALLKFVQLEGAHPLEKSKWEGNYLFPRELFKLVVGGLLSPEEDQSLLLSQFREYLDYDDTRYHTMQAAVDAVARVTGQHPEVPPAFWNNAFTLLSAVSLPRREPTVSSFYVKRAELWDTWKVAHLKEHRRVFQAMWLSFL | |
| 0.02 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79050 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction