missing translation for 'onlineSavingsMsg'
Learn More

Nocodazole, MedChemExpress

Product Code. 16451887
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451887 50 mg 50mg
16436216 100 mg 100mg
16425728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451887 Supplier Medchem Express Supplier No. HY13520
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Nocodazole (Oncodazole) is a rapidly-reversible inhibitor of microtubule. Nocodazole binds to β-tubulin and disrupts microtubule assembly/disassembly dynamics, which prevents mitosis and induces apoptosis in tumor cells. Nocodazole inhibits Bcr-Abl, and activates CRISPR/Cas9.

Nocodazole (Oncodazole) is a rapidly-reversible inhibitor of microtubule. Nocodazole binds to β-tubulin and disrupts microtubule assembly/disassembly dynamics, which prevents mitosis and induces apoptosis in tumor cells. Nocodazole inhibits Bcr-Abl, and activates CRISPR/Cas9.

Specifications

Chemical Name or Material Nocodazole
Physical Form Solid
CAS 31430-18-9
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C14H11N3O3S
For Use With (Application) Cancer-Kinase/protease
Synonym Oncodazole R17934
Solubility Information DMSO : 20 mg/mL (66.37 mM; Need ultrasonic)
SMILES O=C(OC)NC1=NC2=CC=C(C(C3=CC=CS3)=O)C=C2N1
Molecular Weight (g/mol) 301.32
Formula Weight 301.32
Percent Purity 98.63%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.