missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOLA3 Recombinant Rabbit Monoclonal Antibody (JE61-51)
GREENER_CHOICE

Product Code. 17858152
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17858152 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17858152 Supplier Invitrogen™ Supplier No. MA544860

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NOLA3 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

This gene is a member of the H/ACA snoRNPs (small nucleolar ribonucleoproteins) gene family. snoRNPs are involved in various aspects of rRNA processing and modification and have been classified into two families: C/D and H/ACA. The H/ACA snoRNPs also include the DKC1, NOLA1 and NOLA2 proteins. These four H/ACA snoRNP proteins localize to the dense fibrillar components of nucleoli and to coiled (Cajal) bodies in the nucleus. Both 18S rRNA production and rRNA pseudouridylation are impaired if any one of the four proteins is depleted. The four H/ACA snoRNP proteins are also components of the telomerase complex. This gene encodes a protein related to Saccharomyces cerevisiae Nop10p.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOLA3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE61-51
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene NOP10
Gene Accession No. Q9CQS2, Q9NPE3
Gene Alias DKCB1; H/ACA ribonucleoprotein complex subunit 3; homolog of yeast Nop10p; NOLA3; NOP10; NOP10 ribonucleoprotein; NOP10 ribonucleoprotein homolog; NOP10P; nucleolar protein 10; Nucleolar protein family A member 3; nucleolar protein family A, member 3 (H/ACA small nucleolar RNPs); snoRNP protein NOP10
Gene Symbols NOP10
Host Species Rabbit
Immunogen Recombinant protein within human NOP10 aa aa 1-64/64.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55505, 66181
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.