missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 572.00 €
Specifications
| Antigen | NOM1 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18139278
|
Novus Biologicals
NBP2-47352 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18643016
|
Novus Biologicals
NBP2-47352-25ul |
25 μL |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NOM1 Polyclonal specifically detects NOM1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| NOM1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C7orf3, chromosome 7 open reading frame 3, FLJ16401, nucleolar MIF4G domain-containing protein 1, nucleolar protein with MIF4G domain 1, SGD1, SGD1 homolog | |
| NOM1 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 64434 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ITDKSLCGSGEKYIPPHVRQAEETVDFKKKEELERLKKHVKGLLNRLSEPNMASISGQLEELYMAHSRKDMNDTLTSALMGACVTASAMPSRLMMEHVLLVSILHHTVGIEVGAHFLEAVVRKFDAIYKYGSE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title