missing translation for 'onlineSavingsMsg'
Learn More

Nomega-(4-Methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine monohydrate, 95%, Thermo Scientific™

Product Code. 15448456
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15448456

Brand: Thermo Scientific Alfa Aesar H63697.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80745-10-4
Molecular Formula C16H26N4O5S
Molecular Weight (g/mol) 386.467
MDL Number MFCD01862361
InChI Key FUSAEZSSVDNYPO-LBPRGKRZSA-N
Synonym h-arg mtr-oh, h-arg mtr-oh.1/2h2o, s-2-amino-5-3-4-methoxy-2,3,6-trimethylphenyl sulfonyl guanidino pentanoic acid, l-arginyl-4-methoxy-2,3,6-trimethylbenzenesulfonyl, n'-4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine, nomega-4-methoxy-2,3,6-trimethylbenzenesulfonyl-d-arginine, nomega-4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine, amino acid series, arginine mtr-oh, n pound inverted question mark 4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine
PubChem CID 7408178
IUPAC Name (2S)-2-amino-5-[[amino-[(4-methoxy-2,3,6-trimethylphenyl)sulfonylamino]methylidene]amino]pentanoic acid
SMILES CC1=CC(=C(C(=C1S(=O)(=O)NC(=NCCCC(C(=O)O)N)N)C)C)OC

Specifications

Color White
Quantity 1 g
Formula Weight 404.48
Sensitivity Moisture sensitive
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nomega-(4-Methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.