missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Assay Range | 10 to 3160 pg/mL |
| Assay Sensitivity | 2 pg/mL |
| Immunoassay Kit Format | ELISA (Standard) |
| Product Type | ELISA Kit |
| Sample Type | Serum, Plasma, Cell Culture Supernatant |
| Target Species | Non-human Primate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3569 |
| Gene Symbol | IL6 |
| Quantity | 1 Kit (1 Plate) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?