missing translation for 'onlineSavingsMsg'
Learn More

non-muscle heavy chain 10 Myosin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-55146

Product Code. 18291134

  • 483.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

non-muscle heavy chain 10 Myosin Polyclonal specifically detects non-muscle heavy chain 10 Myosin in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

non-muscle heavy chain 10 Myosin
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
Cellular myosin heavy chain, type B, cellular myosin heavy chain, type B type B, heavy polypeptide 10, non-muscle, MGC134913, MGC134914, Myosin heavy chain 10, Myosin heavy chain, non-muscle IIb, myosin heavy chain, nonmuscle type B, myosin, heavy chain 10, non-muscle, myosin-10, near to the ATP binding region, NMMHC II-b, NMMHCB, NMMHC-B, NMMHC-IIB, Non-muscle myosin heavy chain B, nonmuscle myosin heavy chain IIB, Non-muscle myosin heavy chain IIb, nonmuscle myosin heavy chain-B, nonmuscle myosin II heavy chain-B
Rabbit
229 kDa
100 μL
Primary
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
P35580
MYH10
Synthetic peptides corresponding to MYH10(myosin, heavy chain 10, non-muscle) The peptide sequence was selected from the N terminal of MYH10. Peptide sequence WFPKATDKTFVEKLVQEQGSHSKFQKPRQLKDKADFCIIHYAGKVDYKAD.
Affinity purified
RUO
4628
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only