Learn More
Ambion™ Non-Standard Custom Silencer™ siRNA, Custom Size & Purity
Product Code. 15521144
Change view
Click to view available options
Content And Storage:
Room temperature
Unit Size:
Each
Product Code. Content And Storage Unit Size
15521144 Room temperature Each
1 options
Product Code. 15521144 Supplier Ambion™ Supplier No. AM16099
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Non-Standard Custom Silencer™ siRNA, Custom Size & Purity
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description Nonstandard Custom siRNA, Custom Format, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA
Product Type Nonstandard Custom siRNA