missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ NOP9 293T Cell Transient Overexpression Lysate

Product Code. 18540023 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18540023 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18540023 Supplier Novus Biologicals™ Supplier No. H00161424T01
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target NOP9
Synonym C14orf21, chromosome 14 open reading frame 21, KIAA2021, pumilio domain-containing protein C14orf21
Gene Symbol NOP9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.