missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nope Monoclonal Antibody (261123)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA560978
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
| Nope | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9EQS9 | |
| IGDCC4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Nope/IGDCC4, Gly22-His953. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 261123 | |
| Unconjugated | |
| IGDCC4 | |
| amplimer WI-16786, mouse homolog; amplimer WI-18508, mouse homolog; DDM36; hDDM36; Igdcc4; Immunoglobulin superfamily DCC subclass member 4; immunoglobulin superfamily, DCC subclass, member 4; Kiaa1628; likely ortholog of mouse neighbor of Punc E11; LOC57722; neighbor of Punc E11; neighbor of Punc e11 protein; Nope; NOPEFLJ42051; Protein DDM36 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 56741 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction