missing translation for 'onlineSavingsMsg'
Learn More

Nordihydroguaiaretic acid, 95%

Product Code. 10577883 Shop All Thermo Scientific Chemicals Products
Ansicht ändern
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
Packungsgröße:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging Packungsgröße
10577883 1 g Glass bottle 1g
10288983 500 mg Glass bottle 500mg
10280253 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10577883 Lieferant Thermo Scientific Acros Lieferanten-Nr. 376300010
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 500-38-9 | C18H22O4 | 302.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 500-38-9
Molecular Formula C18H22O4
Molecular Weight (g/mol) 302.37
MDL Number MFCD00002206
InChI Key HCZKYJDFEPMADG-UHFFFAOYSA-N
Synonym nordihydroguaiaretic acid, ndga, dihydronorguaiaretic acid, norhydroguaiaretic acid, nordihydroguairaretic acid, norguaiaretic acid, dihydro, 1,2-benzenediol, 4,4'-2,3-dimethyl-1,4-butanediyl bis, dinorguaiaretic acid, dihydro, 4,4'-2,3-dimethyltetramethylene dipyrocatechol, 2,3-bis 3,4-dihydroxyphenylmethyl butane
PubChem CID 4534
ChEBI CHEBI:7625
IUPAC Name 4-[4-(3,4-dihydroxyphenyl)-2,3-dimethylbutyl]benzene-1,2-diol
SMILES CC(CC1=CC(=C(C=C1)O)O)C(C)CC2=CC(=C(C=C2)O)O

Spezifikation

CAS Min % 94.0
CAS Max % 100.0
Melting Point 184°C to 189°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula [(OH)2C6H3CH2CH(CH3)]2
Quantity 1 g
Beilstein 6,IV,7771
Merck Index 15,6782
Solubility Information Solubility in water: slightly soluble in hot water. Other solubilities: soluble in methanol,ethanol,acetone,slightly soluble in chloroform,insoluble in petroleum ether
Formula Weight 302.37
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nordihydroguaiaretic acid
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Nordihydroguaiaretic acid
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.