missing translation for 'onlineSavingsMsg'
Learn More

Norepinephrine, MedChemExpress

Product Code. 16441696
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441696 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441696 Supplier Medchem Express Supplier No. HY13715
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Norepinephrine (Levarterenol; L-Noradrenaline) is a potent adrenergic receptor (AR) agonist. Norepinephrine activates α1, α2, β1 receptors.

Norepinephrine (Levarterenol

Specifications

Chemical Name or Material Norepinephrine
CAS 51-41-2
Color Khaki
Purity Grade Notes Research
Molecular Formula C8H11NO3
Quantity 500 mg
Synonym Levarterenol L-Noradrenaline
Solubility Information DMSO : 25 mg/mL (147.77 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC1=CC=C([C@@H](O)CN)C=C1O
Molecular Weight (g/mol) 169.18
Formula Weight 169.18
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.