missing translation for 'onlineSavingsMsg'
Learn More

Norflurazon-desmethyl 10 μg/mL in Ethyl acetate, Dr. Ehrenstorfer

Product Code. 30326417
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326417 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326417 Supplier Dr. Ehrenstorfer Supplier No. DRELA15651000EA
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Norflurazon-desmethyl
CAS 23576-24-1
Analyte or Component Names Norflurazon-desmethyl
Shipping Condition Room Temperature
Molecular Formula C11 H7 Cl F3 N3 O
Concentration 10 μg/mL
Certifications/Compliance ISO 17034
Synonym 3(2H)-Pyridazinone, 5-amino-4-chloro-2-[3-(trifluoromethyl)phenyl]-, 3(2H)-Pyridazinone, 5-amino-4-chloro-2-(α,α,α-trifluoro-m-tolyl)- (8CI), 5-Amino-4-chloro-2-[3-(trifluoromethyl)phenyl]-3(2H)-pyridazinone, 4-Chloro-5-amino-2-(α,α,α-trifluoro-m-tolyl)-3(2H)-pyridazinone, Desmethylnorflurazon, SAN 9774, Norflurazon-desmethyl
InChI Formula InChI=1S/C11H7ClF3N3O/c12-9-8(16)5-17-18(10(9)19)7-3-1-2-6(4-7)11(13,14)15/h1-5H,16H2
Solution Type Single Solution
SMILES NC1=C(Cl)C(=O)N(N=C1)c2cccc(c2)C(F)(F)F
IUPAC Name 5-amino-4-chloro-2-[3-(trifluoromethyl)phenyl]pyridazin-3-one
Solvent or Matrix Ethyl Acetate
Industry Type Environmental Testing
Molecular Weight (g/mol) 289.64
Quantity 1 mL
Formula Weight 289.023 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.