missing translation for 'onlineSavingsMsg'
Learn More

(-)-Normacromerine, TRC

Product Code. 30410509
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30410509 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410509 Supplier Toronto Research Chemicals Supplier No. TRCN717000500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41787-64-8
Molecular Formula C11H17NO3
Molecular Weight (g/mol) 211.26
Synonym (alphaR)-3,4-Dimethoxy-alpha-[(methylamino)methyl]benzenemethanol, Normacromerine, (R)-3,4-Dimethoxy-alpha-[(methylamino)methyl]benzenemethanol
IUPAC Name (1R)-1-(3,4-dimethoxyphenyl)-2-(methylamino)ethanol
SMILES CNC[C@H](O)c1ccc(OC)c(OC)c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H17NO3/c1-12-7-9(13)8-4-5-10(14-2)11(6-8)15-3/h4-6,9,12-13H,7H2,1-3H3/t9-/m0/s1
Formula Weight 211.1208
Chemical Name or Material (-)-Normacromerine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.