missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Normal Rabbit Serum

Product Code. 18786133 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18786133 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18786133 Supplier R&D Systems™ Supplier No. NBP171681
  • ONLINE EXCLUSIVE PRICE
    453.23 €
    333.90 € / 5mL
    Save 119.33 €
    26% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Applications: Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Control, Immunocytochemistry, Block/Neutralize

  • Host: Rabbit
  • Contains 0.1% Sodium Azide as presevative
Applications: 

Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin

Recommended Storage

Store at 4°C for short term and -20°C for long term

TRUSTED_SUSTAINABILITY

Specifications

Species Rabbit
Product Type Rabbit Serum
Quantity 5 mL
For Use With (Application) Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Control, Block/Neutralize, Fluorescence Imaging
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.