missing translation for 'onlineSavingsMsg'
Learn More

Norverapamil hydrochloride, MedChemExpress

Product Code. 16485887
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485887 5 mg 5mg
16470036 2 mg 2mg
16462646 25 mg 25mg
16443158 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485887 Supplier Medchem Express Supplier No. HY100750
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Norverapamil hydrochloride ((±)-Norverapamil hydrochloride), an N-demethylated metabolite of Verapamil, is a L-type calcium channel blocker and a P-glycoprotein (P-gp) function inhibitor.

Norverapamil hydrochloride ((±)-Norverapamil hydrochloride), an N-demethylated metabolite of Verapamil, is a L-type calcium channel blocker and a P-glycoprotein (P-gp) function inhibitor.

Specifications

Chemical Name or Material Norverapamil hydrochloride
CAS 67812-42-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H37ClN2O4
Quantity 5 mg
Synonym (±)-Norverapamil hydrochlorideD591 hydrochloride
Solubility Information H2O : ≥ 50 mg/mL (104.81 mM) ∣DMSO : ≥ 31 mg/mL (64.98 mM)
SMILES N#CC(C(C)C)(CCCNCCC1=CC=C(OC)C(OC)=C1)C2=CC=C(OC)C(OC)=C2.[H]Cl
Molecular Weight (g/mol) 477.04
Formula Weight 477.04
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.