missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOS1AP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-38151
This item is not returnable.
View return policy
Description
NOS1AP Polyclonal specifically detects NOS1AP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| NOS1AP | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| O75052 | |
| NOS1AP | |
| This antibody was developed against a recombinant protein corresponding to amino acids: HTGSKVSHPQEPMLTASPRMLLPSSSSKPPGLGTETPLSTHHQMQLLQQLLQQQQQQTQVAVAQVHLLKDQLAAEAAARLEAQARVHQLLLQNKDMLQHISLLVK | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 6330408P19Rik, CAPONMGC138500, carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase protein, C-terminal PDZ ligand of neuronal nitric oxide synthase protein, KIAA0464C-terminal PDZ domain ligand of neuronal nitric oxide synthase (CAPON), ligand of neuronal nitric oxide synthase with carboxyl-terminal PDZ domain, nitric oxide synthase 1 (neuronal) adaptor protein, Nitric oxide synthase 1 adaptor protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9722 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction