missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 20X Bolt™ MOPS SDS Running Buffer

Product Code. 13226499
Change view
Click to view available options
Quantity:
500 mL
5 L
Unit Size:
5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13226499 500 mL 500mL
13236499 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13226499 Supplier Invitrogen™ Supplier No. B0001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimized for use with Bolt™ Bis-Tris Plus gels, and are available in a variety of formats

About Bolt™ Bis-Tris Plus Gels
Bolt™ Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of your small- to medium-sized proteins under denaturing conditions. Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt™ gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt™ gels to obtain optimal results for your day-to-day protein separation needs. Bolt™ Bis-Tris Plus gels come in four gel types and multiple well formats. A Mini Gel Tank is required for use of all Bolt™ gels.

Bolt™ gel run times may vary depending upon the type of running buffer used.

Order Info

Shelf-life is 6 months, when stored properly.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Running Buffer
Concentration 20X
For Use With (Equipment) Bolt
Physical Form Liquid
Product Line Bolt
Quantity 500 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.