Learn More
Invitrogen™ NuPAGE™ MES SDS Running Buffer (20X)
Description
Includes
NuPAGE MES SDS Running Buffer (20X)
Requires
The buffers are provided as a concentrated solution and require simple dilution with deionized water before use.
NuPAGE MES SDS Running Buffer (20X) is formulated for running proteins on NuPAGE Novex Bis-Tris gels only. NuPAGE MES SDS Running Buffer is recommended for separating small- to medium-sized proteins.
- Use of MES buffer allows proteins to run faster
- Ensures high-quality, consistent electrophoresis results
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | Running Buffer |
| Concentration | 20X |
| pH | 7.3 |
| Physical Form | Liquid |
| Product Line | NuPAGE |
| Quantity | 5 L |
| Formulation | 1X: 50 mM MES, 50 mM Tris Base, 0.1% SDS, 1 mM EDTA, pH 7.3 |
Frequently Asked Questions (FAQs)
DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.
Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.
While they are both Bis-Tris based gels, the chemistries are very different since Bolt gels are optimized for western blotting. Another key difference is the wedge well design of the Bolt gels, which allows larger sample volumes to be loaded.
The neutral operating pH of the NuPAGE Gels and buffers provides following advantages over the Laemmli system:
-Longer shelf life of 8-12 months due to improved gel stability
-Improved protein stability during electrophoresis at neutral pH resulting in sharper band resolution and accurate results (Moos et al, 1998)
-Complete reduction of disulfides under mild heating conditions (70 degrees C for 10 min) and absence of cleavage of asp-pro bonds using the NuPAGE LDS Sample buffer (pH > 7.0 at 70 degrees C)
-Reduced state of the proteins maintained during electrophoresis and blotting of the proteins by the NuPAGE Antioxidant
Please refer to the following paper: Moos M Jr, Nguyen NY, Liu TY (1988) Reproducible High Yield Sequencing of Proteins Electrophoretically Separated and Transferred to an Inert Support. J Biol Chem 263:6005-6008.
Both the NuPAGE MES SDS Running Buffer (20X) (Cat. No. NP0002) and the 20X Bolt MES SDS Running Buffer (Cat. No. B0002) have the exact same composition: 50 mM MES, 50 mM Tris Base, 0.1% SDS, 1 mM EDTA, pH 7.3.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.