missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex™ Tris-Glycine Native Sample Buffer (2X)
Description
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Chemical Name or Material | Sample Loading Buffer |
| Concentration | 2X |
| For Use With (Equipment) | Mini Precast Protein Gels |
| pH | 8.6 |
| Physical Form | Liquid |
| Product Line | Novex |
| Quantity | 20 mL |
| Formulation | Glycerol 56-81-5 (20-30) |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?