missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Novobiocin ReadyMade™ Solution

Product Code. 30044032
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
10 x 1 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044032 10 x 1 mL 10mL
30044018 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30044032 Supplier TOKU-E™ Supplier No. N08910x1ML
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Novobiocin ReadyMade™ Solution is provided as a sterile-filtered solution of Novobiocin Sodium in water.

  • CAS Number: 1476-53-5
  • Molecular Formula: C31H35N2NaO11
  • Molecular Weight: 634.61 g/mol
  • Storage Conditions: -20°C, Protect from light
  • Form: Liquid
  • Source: Streptomyces spp.
  • Spectrum: Novobiocin Sodium targets primarily Gram-positive organisms and is commonly used to differentiate between coagulase-negative Staphylococcus species (all but S. aureus and a few others), with S. saprophyticus being resistant to novobiocin in culture
  • Mechanism of Action: Aminocoumarin antibiotics target DNA-gyrase, an enzyme which relieves strain as DNA unwinds during replication and transcription. It also inhibits mammalian topoisomerase, both are ATP-dependent enzymes involved with cell division

Specifications

Chemical Name or Material Novobiocin Sodium
Physical Form Liquid
CAS 1476-53-5
Source Streptomyces spp.
Packaging 2 mL plastic vial
Sterile Filtered Yes, 0.22 μm
Sterility Sterile
Color Yellow
Concentration 100 to 105 mg/mL
Quantity 10 x 1 mL
Purity Grade Notes >90% (HPLC)
Assay Percent Range >850 μg/mg
Assay Percent Notes Novobiocin Sodium, Dried Basis
Molecular Formula C31H35N2NaO11
Product Line ReadyMade™ Solutions
Shipping Condition Ice Pack
IUPAC Name sodium;7-[(2R,3R,4S,5R)-4-carbamoyloxy-3-hydroxy-5-methoxy-6,6-dimethyloxan-2-yl]oxy-3-[[4-hydroxy-3-(3-methylbut-2-enyl)benzoyl]amino]-8-methyl-2-oxochromen-4-olate
Molecular Weight (g/mol) 634.61 g/mol
Formula Weight 634.61 amu
Percent Purity >90%
Grade Antibiotic
Concentration or Composition (by Analyte or Components) Novobiocin Sodium ≥5 to <10%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.