missing translation for 'onlineSavingsMsg'
Learn More

Novobiocin Sodium Salt, TRC

Product Code. 30009873
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009873 10 g 10g
30009797 5 g 5g
30009789 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009873 Supplier Toronto Research Chemicals Supplier No. TRCN88850010G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1476-53-5
Molecular Formula C31 H35 N2 O11 . Na
Molecular Weight (g/mol) 634.61
Synonym Benzamide, N-[7-[[3-O-(aminocarbonyl)-6-deoxy-5-C-methyl-4-O-methyl-α-L-lyxo-hexopyranosyl]oxy]-4-hydroxy-8-methyl-2-oxo-2H-1-benzopyran-3-yl]-4-hydroxy-3-(3-methyl-2-buten-1-yl)-, sodium salt (1:1) (ACI), Benzamide, N-[7-[[3-O-(aminocarbonyl)-6-deoxy-5-C-methyl-4-O-methyl-α-L-lyxo-hexopyranosyl]oxy]-4-hydroxy-8-methyl-2-oxo-2H-1-benzopyran-3-yl]-4-hydroxy-3-(3-methyl-2-butenyl)-, monosodium salt (9CI), Novobiocin, monosodium salt (8CI), Novobiocin, sodium deriv. (6CI, 7CI), Albamycin, Albamycin (capsule), Albamycin sodium, Cathomycin sodium, Cathomycin sodium lyovac, Monosodium novobiocin, Novobiocin monosodium, Novobiocin sodium, Novobiocin sodium salt, NSC 2382, Sodium albamycin, Sodium novobiocin, Vulcamycin
IUPAC Name sodium;7-[(2R,3R,4S,5R)-4-carbamoyloxy-3-hydroxy-5-methoxy-6,6-dimethyloxan-2-yl]oxy-3-[[4-hydroxy-3-(3-methylbut-2-enyl)benzoyl]amino]-8-methyl-2-oxochromen-4-olate
SMILES [Na+].CO[C@@H]1[C@@H](OC(=O)N)[C@@H](O)[C@H](Oc2ccc3C(=C(NC(=O)c4ccc(O)c(CC=C(C)C)c4)C(=O)Oc3c2C)[O-])OC1(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C31H36N2O11.Na/c1-14(2)7-8-16-13-17(9-11-19(16)34)27(37)33-21-22(35)18-10-12-20(15(3)24(18)42-28(21)38)41-29-23(36)25(43-30(32)39)26(40-6)31(4,5)44-29;/h7,9-13,23,25-26,29,34-36H,8H2,1-6H3,(H2,32,39)(H,33,37);/q;+1/p-1/t23-,25+,26-,29-;/m1./s1
Formula Weight 634.2139
Percent Purity >95%
Chemical Name or Material Novobiocin sodium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.