Learn More
Novus Biologicals™ Fascin Protein
Product Code. p-5354741 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.5mg
Product Code. 18747553
Brand: Novus Biologicals™ NBP1453130.1mg
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-493 of Human Fascin The Recombinant Human Fascin Protein is derived from E. coli. The Recombinant Human Fascin Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.1 mg