missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HeLa Whole Cell Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Description
The HeLa Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Reproductive |
| Quantity | 0.1 mg |
| Storage Buffer | Standard RIPA buffer including protease inhibitors and 0.1% SDS diluted in 1X lithium dodecyl sulfate (LDS) sample buffer with bromophenol blue and 40-70% glycerol, pH 8.4. Denaturing agents have not been added. |
| Purity | Protein |
| Cellular Fraction | Whole |
| Content And Storage | Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction