missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ HepG2 Whole Cell Lysate

Product Code. 18716463 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18716463

Brand: Novus Biologicals™ NBP142569

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: Western Blot, Immunoprecipitation, SDS-Page

The HepG2 Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation, SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Immunoprecipitation, SDS-Page, Western Blot
Tissue Liver
Quantity 1 mg
Storage Buffer Standard RIPA buffer including protease and phosphatase inhibitors and 0.1% SDS diluted in 4X Laemmli Sample Buffer [62.5mM Tris HCl, 1% LDS, 10% Glycerol, 0.005% bromophenol blue, pH 6.8] BME has not been added.
Test Specificity HepG2
Cellular Fraction Whole
Host Species Human
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.