missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Histone H4 [Monomethyl Arg3] Antibody Blocking Peptide
Shop All Bio Techne ProductsSpecifications
Specifications
| Gene ID (Entrez) | 121504 |
| Species | Human |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Lyophilized. Reconstitute in 40 ul of diH20. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB21-2011. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 |
| Gene Symbol | HIST4H4 |
| Product Type | Histone H4 |
| Quantity | 0.04 mg |
| Regulatory Status | RUO - research use only |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?