Learn More
Novus Biologicals™ HIV-1 p24 Protein
Product Code. 18710974 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.5mg
Product Code. 18710974
Brand: Novus Biologicals™ NBP1460360.5MG
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 155-321 of HIV-1 HIV-1 p24 The Recombinant Virus HIV-1 p24 Protein is derived from E. coli. The Recombinant Virus HIV-1 p24 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.5 mg