Learn More
Novus Biologicals™ HPRT Protein
Product Code. p-5354853 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.10mg
0.5mg
Product Code. 18766013
Brand: Novus Biologicals™ NBP1452510.5MG
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-218 of Human HPRT The Recombinant Human HPRT Protein is derived from E. coli. The Recombinant Human HPRT Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.5 mg