missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 20-129 of Human Substance P The Recombinant Human Substance P Protein is derived from E. coli. The Recombinant Human Substance P Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Purification Method | SDS-PAGE |
| Quantity | 0.1 mg |
| Storage Requirements | −20°C or −80°C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Gene Alias | Hs0.2563, NK2, NKNA, NPK, PPT, protachykinin-1, TAC1, TAC2 |
| Conjugate | Unconjugated |
| Species | Human |
| Recombinant | Recombinant |
| Product Line | Novus |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?