missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IkB-alpha Protein

Product Code. 18253132 Shop All Bio Techne Products
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
This item is not returnable. View return policy

Product Code. 18253132

Brand: Novus Biologicals™ NBP172500250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-317 of Human IkB-alpha The Recombinant Human IkB-alpha Protein is derived from E. coli. The Recombinant Human IkB-alpha Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 20% glycerol, 0.1M NaCl
Quantity 250 μg
Source Human
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name IkB-alpha
Conjugate Unconjugated
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.