missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MAP3K12 Antibody Blocking Peptide

Product Code. 18264972 Shop All Bio Techne Products
Click to view available options
Quantity:
0.25 mg
Unit Size:
0.25mg
This item is not returnable. View return policy

Product Code. 18264972

Brand: Novus Biologicals™ NBP146063PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

Antigenic Blocking Peptide corresponding to amino acids 260 - 320 of human MAP3K12. The MAP3K12 Blocking Peptide is derived from Synthetic. The MAP3K12 Blocking Peptide has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 7786
Species Human
Purity >85%, by HPLC
Accession Number Q12852
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation Sodium Bicarbonate buffer, pH 8.00 containing up to 10% DMSO
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-46063. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias DLKEC 2.7.11.25, Dual leucine zipper bearing kinase, EC 2.7.11, Leucine-zipper protein kinase, MAPK-upstream kinase, MEKK12, mitogen-activated protein kinase kinase kinase 12, Mixed lineage kinase, MUKdual leucine zipper kinase DLK, protein kinase MUK, ZPKleucine zipper protein kinase, ZPKP1
Gene Symbol MAP3K12
Product Type MAP3K12
Quantity 0.25 mg
Regulatory Status RUO - research use only
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.