missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ NIH 3T3 Whole Cell Lysate

Product Code. 18727053 Shop All Bio Techne Products
Click to view available options
:
0.1mL
This item is not returnable. View return policy

Product Code. 18727053

Brand: Novus Biologicals™ NB800PC8

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
The NIH 3T3 Whole Cell Lysate is derived from mouse. It has been validated for the following applications: Western Blot, SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Tissue Bone/Cartilage
Quantity 0.1 mL
Storage Buffer Standard RIPA buffer including protease and phosphatase inhibitors and 0.1% SDS diluted in 1X lithium dodecyl sulfate (LDS) sample buffer with bromophenol blue and 40-70% glycerol, pH 8.4. BME has not been added.
Purity Protein
Test Specificity NIH/3T3 whole cell lysate (Positive control)
Cellular Fraction Whole
Content And Storage -80°C. Avoid freeze-thaw cycles.

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.