missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Protein A Native Protein

Product Code. 18027416 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18027416 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18027416 Supplier Novus Biologicals™ Supplier No. NBP197240
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Western Blot, Immunoprecipitation

Native Protein A. The Protein A Native Protein has been validated for the following applications: Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein A Protein
Quantity 2 mL
Concentration 6 mg/mL
For Use With (Application) Western Blot, Immunoprecipitation
Gene ID (Entrez) 3919448
Gene Symbol SAOUHSC_00069
Protein Subtype Protein A
Content And Storage Store at 4°. Do not freeze.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.