missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Raw 264.7 Whole Cell - T0901317 treated Whole Cell Lysate
Shop All Bio Techne Products
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1 mL
missing translation for 'unitSize'
0.10mL
Description
The Raw 264.7 Whole Cell Lysate is derived from mouse. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Blood Lymph |
| Quantity | 0.1 mL |
| Storage Buffer | Standard RIPA buffer including protease and phosphatase inhibitors and 0.1% SDS diluted in 1X lithium dodecyl sulfate (LDS) sample buffer with bromophenol blue and 40-70% glycerol, pH 8.4. BME has not been added. |
| Cellular Fraction | Whole |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Target | Raw 264.7 Whole Cell - T0901317 treated |
| Protein State | Denatured |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction