Learn More
Novus Biologicals™ S100A1 Protein
Product Code. p-5355298 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.5mg
Product Code. 18741044
Brand: Novus Biologicals™ NBP1723350.1mg
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-94 of Mouse S100A1 The Recombinant Mouse S100A1 Protein is derived from E. coli. The Recombinant Mouse S100A1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.1 mg