Learn More
Novus Biologicals™ S100A8 Protein
Product Code. p-5355311 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.10mg
0.5mg
Product Code. 18771554
Brand: Novus Biologicals™ NBP1723310.1mg
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-89 of Mouse S100A8 The Recombinant Mouse S100A8 Protein is derived from E. coli. The Recombinant Mouse S100A8 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.1 mg