missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SD OLLAS/DYKDDDDK Vector

Product Code. 18765143 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.01 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18765143 0.01 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18765143 Supplier Novus Biologicals™ Supplier No. NBA107085

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

pCMV-SD OLLAS / DYKDDDDK Vector for expression of secretory protein with N-terminally FLAG(DYKDDDDK) and OLLAS dual tandem tagged

The pCMV-NS-OLLAS vector provides a convenient vector to express recombinant proteins under control of the CMV promoter with both an N-terminal OLLAS tag and a DYKDDDDK tag, protein will be secreted from the cell due to the inclusion of a signal peptide. Insert protein of interest into the vector in frame using the Xba I and EcoR I/Not I restriction sites. Bacterial selection can be performed using Kanamycin, while Neomycin resistance can be used for screening mammalian transfectants.

Recommended Storage

Store at -20°C. Avoid freeze-thaw cycles.

TRUSTED_SUSTAINABILITY

Specifications

Storage Buffer 5mM Tris pH 8
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 0.01 mg
Vector SD OLLAS/DYKDDDDK Vector
For Use With (Application) For expression of secretory protein with N-terminally FLAG(DYKDDDDK) and OLLAS dual tandem tagged
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.