Learn More
Novus Biologicals™ Chromata ChIP Histone H3 [ac Lys9, ac Lys14] Kit (ChIP)
Shop All Bio Techne ProductsDescription
Includes
10x glycine, IP dilution buffer, H3 K4Me3 positive control antibody, Protein A/G magnetic beads, IP wash buffer 1, IP wash buffer 2, IP wash buffer 3, IP wash buffer 4, IP elution buffer, 5M NaCL, Proteinase K, Purification columns, Purification column Tubes, DNA binding buffer, DNA wash buffer, DNA elution buffer, Human RPL30 positive control primer pair
Kit contains all the necessary components to perform 25 Immunoprecipitation experiments including a positive control antibody. This ChIP kit is specific for Histone H3 acetylated Lysine 9 / Lysine 14.
Spécification
Spécification
| Kit Contents | 10x glycine, IP dilution buffer, H3 K4Me3 positive control antibody, Protein A/G magnetic beads, IP wash buffer 1, IP wash buffer 2, IP wash buffer 3, IP wash buffer 4, IP elution buffer, 5M NaCL, Proteinase K, Purification columns, Purification column Tubes, DNA binding buffer, DNA wash buffer, DNA elution buffer, Human RPL30 positive control primer pair |
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Quantity | 25 Immunoprecipitations |
| For Use With (Application) | Chromatin Immunoprecipitation of DNA from fixed Hela cells alongside a no antibody (No Ab) control |
| Target | Chromata ChIP Histone H3 |
| Type | ChIP Histone H3 |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.